Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium 2,4-pentanedionate hydrate, 95%

Click to view available options
Quantity:
50g
250g
1000g
86891-03-4
C5H9NaO3
140.114
MFCD00078034
UKDRZWCRRJOZNR-CZEFNJPISA-M
sodium 2,4-pentanedionate,sodium 2e-4-oxopent-2-en-2-olate hydrate
90476915
sodium;(E)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].O.[Na+]
86891-03-4
C5H9NaO3
140.114
MFCD00078034
UKDRZWCRRJOZNR-CZEFNJPISA-M
sodium 2,4-pentanedionate,sodium 2e-4-oxopent-2-en-2-olate hydrate
90476915
sodium;(E)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].O.[Na+]
Applications
Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, water/ moisture, acids.
Melting Point 228°C to 230°C
Quantity 1000g
Formula Weight 122.11 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Sodium 2,4-pentanedionate hydrate

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA