Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Amino-5-nitrobenzenesulfonate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
30693-53-9
C6H5N2NaO5S
240.165
MFCD00068347
VTMRGLXTSDVXAE-UHFFFAOYSA-M
2-Amino-5-nitrobenzenesulfonic Acid Sodium Salt, 4-Nitroaniline-2-sulfonic Acid Sodium Salt, Sodium 4-Nitroaniline-2-sulfonate
121742
sodium;2-amino-5-nitrobenzenesulfonate
C1=CC(=C(C=C1[N+](=O)[O-])S(=O)(=O)[O-])N.[Na+]
30693-53-9
C6H5N2NaO5S
240.165
MFCD00068347
VTMRGLXTSDVXAE-UHFFFAOYSA-M
2-Amino-5-nitrobenzenesulfonic Acid Sodium Salt, 4-Nitroaniline-2-sulfonic Acid Sodium Salt, Sodium 4-Nitroaniline-2-sulfonate
121742
sodium;2-amino-5-nitrobenzenesulfonate
C1=CC(=C(C=C1[N+](=O)[O-])S(=O)(=O)[O-])N.[Na+]
Quantity 500g
Color Yellow
Formula Weight 240.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Sodium 2-Amino-5-nitrobenzenesulfonate

EINECSNumber : (3)-2079

TSCA : Yes