Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

Catalog Number AAB249600I
Click to view available options
Quantity:
250 g
1000 g
5000 g
19766-89-3
C8H15NaO2
166.20
MFCD00014007
VYPDUQYOLCLEGS-UHFFFAOYNA-M
sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate
23672331
sodium;2-ethylhexanoate
[Na+].CCCCC(CC)C([O-])=O
19766-89-3
C8H15NaO2
166.20
MFCD00014007
VYPDUQYOLCLEGS-UHFFFAOYNA-M
sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate
23672331
sodium;2-ethylhexanoate
[Na+].CCCCC(CC)C([O-])=O

Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Odor Odorless
Quantity 5000 g
Solubility Information Soluble in water.
Formula Weight 166.2
Percent Purity 97%
Chemical Name or Material Sodium 2-ethylhexanoate
Hazard Category H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 243-283-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit