Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium 4-(tert-butylcarbonyloxy)benzenesulfonate, 95%

Click to view available options
Quantity:
1g
5g
25g
188114-91-2
C11H13NaO5S
280.27
MFCD09859793
YJWRXNUCTLGRAC-UHFFFAOYSA-M
sodium 4-pivaloyloxy benzenesulfonate,sodium 4-tert-butylcarbonyloxy-benzensulfonate,sodium 4-t-butylcarbonyloxy-benzensulfonate,sodium 4-t-butylcarbonyloxy benzensulfonate,sodium 4-2,2-dimethylpropanoyl oxy benzenesulfonate,sodium 4-t-butylcarbonyloxy-benzensulf...,4-pivaloyloxy benzenesulfonic acid sodium salt,2,2-dimethylpropanoic acid 4-sulfophenyl ester sodium salt,sodium 4-2,2-dimethylpropanoyl oxy benzene-1-sulfonate,propanoic acid, 2,2-dimethyl-, 4-sulfophenyl ester, sodium salt 1:1
23717480
sodium;4-(2,2-dimethylpropanoyloxy)benzenesulfonate
CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
188114-91-2
C11H13NaO5S
280.27
MFCD09859793
YJWRXNUCTLGRAC-UHFFFAOYSA-M
sodium 4-pivaloyloxy benzenesulfonate,sodium 4-tert-butylcarbonyloxy-benzensulfonate,sodium 4-t-butylcarbonyloxy-benzensulfonate,sodium 4-t-butylcarbonyloxy benzensulfonate,sodium 4-2,2-dimethylpropanoyl oxy benzenesulfonate,sodium 4-t-butylcarbonyloxy-benzensulf...,4-pivaloyloxy benzenesulfonic acid sodium salt,2,2-dimethylpropanoic acid 4-sulfophenyl ester sodium salt,sodium 4-2,2-dimethylpropanoyl oxy benzene-1-sulfonate,propanoic acid, 2,2-dimethyl-, 4-sulfophenyl ester, sodium salt 1:1
23717480
sodium;4-(2,2-dimethylpropanoyloxy)benzenesulfonate
CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
Quantity 1g
Formula Weight 280.27
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Sodium 4-(tert-butylcarbonyloxy)benzenesulfonate