Learn More
Sodium 99.8%, oiled sticks, wrapped in aluminium foil, ACROS Organics™
Specifications
Quantity | 250g |
Calcium (Ca) | 400ppm max. |
Assay Percent Range | 99.8% |
Linear Formula | Na |
Merck Index | 15, 8708 |
Formula Weight | 23.99 |
Physical Form | Sticks |
Percent Purity | 99.8% |
Potassium (K) | 300ppm max. |
Sodium (Na) | 99.75% min. |
Show More |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger