Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium AA Standard, 1mL = 1mg Na (1,000ppm Na) in 3% HCl, Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
7647-14-5,7732-18-5,7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]
7647-14-5,7732-18-5,7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]
Concentration or Composition Notes 1,000ppm Na
CAS Min % 95.0,3.05,0.22
CAS Max % 99.0,3.18,0.23
Physical Form Liquid
Packaging Poly natural
Equivalence 1 mL = 1 mg Na
Color Colorless
Quantity 100mL
Chemical Name or Material Sodium AA Standard