Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium AA Standard, 1000 ppm Na in 3% HNO3, Ricca Chemical
SDP

Catalog Number p-3255654
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
497-19-8, 7697-37-2, 7732-18-5
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
497-19-8, 7697-37-2, 7732-18-5
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
Physical Form Liquid
CAS Min % 93.0, 4.59, 0.21
CAS Max % 97.0, 4.77, 0.22
Packaging Natural LDPE Bottle
Equivalence 1 mL = 1 mg Na
Color Colorless
Quantity 100 mL
Concentration or Composition Notes 1,000ppm Na
Chemical Name or Material Sodium AA Standard
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit