Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium acetate, ≥99%, anhydrous, Molecular Biology Reagent

Click to view available options
Quantity:
100g
500g
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
Applications
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Merck Index 14,8571
Formula Weight 82.03
Physical Form Powder
Percent Purity ≥99%
Sensitivity Hygroscopic
Color White
Melting Point 324°C
pH 7.5 to 9
Quantity 500g
Chemical Name or Material Sodium acetate, Anhydrous, Molecular Biology Reagent

RTECSNumber : AJ4300010

TSCA : TSCA