Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
127-09-3
C2H3NaO2
82.034
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]
127-09-3
C2H3NaO2
82.034
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech
517045
CHEBI:32954
sodium;acetate
CC(=O)[O-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 82.03
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.53kg
Quantity 2.5kg
Chemical Name or Material Sodium Acetate, Anhydrous