Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate (Colorless-to-white Crystals), Fisher BioReagents

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]

Sodium Acetate Trihydrate (Colorless-to-white Crystals)

The trihydrate form of Sodium Acetate can be used for many of the same applications as the anhydrous form.
Linear Formula CH3COONa·3H2O
Formula Weight 136.08
Physical Form Solid
Percent Purity 99.0 to 101.0%
Calcium (Ca) 0.005% max.
Chloride 0.001% max.
Density 1.45g/cm³
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.005% max.
Iron (Fe) 5ppm max.
Magnesium (Mg) 0.002% max.
Packaging Poly Bottle
Phosphate 5ppm max.
Potassium (K) 0.005% max.
Substances Reducing Permanganate Pass Test
Sulfate 0.002% max.
Color White
Melting Point 58°C
pH 8 to 9.5
Quantity 500g
Chemical Name or Material Sodium Acetate Trihydrate
Show More Show Less

Recommended Storage : RT