Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Trihydrate, Crystal, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]
Formula Weight 136.08
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.45kg
Quantity 2.5kg
Chemical Name or Material Sodium Acetate, Trihydrate
Grade FCC/USP