Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Trihydrate, Granular, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
100 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6131-90-4
C2H9NaO5
136.08
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate
O.O.O.[Na+].CC([O-])=O
6131-90-4
C2H9NaO5
136.08
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate
O.O.O.[Na+].CC([O-])=O

SO105, 6131-90-4, NaC2H3O2•3H2O

Spectrum™ Sodium Acetate, Trihydrate, Granular, USP is used as a source of sodium ions in solutions for dialysis and as a systemic and urinary alkalizer, diuretic, and expectorant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 99 to 101%
Grade USP
Insoluble Matter 0.0005
Loss on Drying 38-41%
Packaging Fiber Drum
Sulfate 5e-005
pH 7.5 9.2
Quantity 100 kg