Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium acetate trihydrate, Honeywell
Click to view available options
Quantity:
250g
500g
1kg
2kg
5kg
Description
- ≥95% for general laboratory use.
Specifications
Linear Formula | CH3COONa · 3H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3732037 |
Formula Weight | 136.08g/mol |
Percent Purity | ≥99% |
Ignition Point | 1112°F |
Quantity | 2kg |
Chemical Name or Material | Sodium acetate trihydrate |
Safety and Handling
RTECSNumber : AJ4580000