Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium acetylide, 18 wt.% slurry in xylene, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1066-26-8,1330-20-7
C2HNa
48.02
MFCD00008568
SFDZETWZUCDYMD-UHFFFAOYSA-N
sodium acetylide,sodium ethynide,ethynyl sodium,sodium acetylide suspension,sodium acetylide na c2h,sodium ethyne,monosodium acetylide,slbnqjagykqcki-uhfffaoysa-n,sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide
2733336
CHEBI:55387
sodium;ethyne
C#[C-].[Na+]
1066-26-8,1330-20-7
C2HNa
48.02
MFCD00008568
SFDZETWZUCDYMD-UHFFFAOYSA-N
sodium acetylide,sodium ethynide,ethynyl sodium,sodium acetylide suspension,sodium acetylide na c2h,sodium ethyne,monosodium acetylide,slbnqjagykqcki-uhfffaoysa-n,sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide
2733336
CHEBI:55387
sodium;ethyne
C#[C-].[Na+]
Color Brown to Yellow
Density 0.9360g/mL
Quantity 100g
Flash Point 25°C
Packaging Glass bottle
Specific Gravity 0.936
Linear Formula HC≡CNa
Solubility Information Solubility in water: reacts.
Formula Weight 48.02
Physical Form Slurry
Percent Purity 17.0 to 22.0 wt%
Chemical Name or Material Sodium acetylide, 18wt% slurry in xylene
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
May be fatal if swallowed and enter

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Brush off loose particles from skin.
Immerse

GHS Signal Word: Danger

EINECSNumber : 213-908-9