Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acid Pyrophosphate, FCC, 93-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7758-16-9
Na4O7P2
265.90
FQENQNTWSFEDLI-UHFFFAOYSA-J
tetrasodium (phosphonatooxy)phosphonate
[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
7758-16-9
Na4O7P2
265.90
FQENQNTWSFEDLI-UHFFFAOYSA-J
tetrasodium (phosphonatooxy)phosphonate
[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

S1099, 7758-16-9, Na2H2P2O7

Spectrum™ Sodium Acid Pyrophosphate, FCC is leavening agent found in baking powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 93 to 100.5%
Fluoride 0.005
Insoluble Matter 0.01
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Quantity 12 kg