Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium aluminium hydride, 93%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
13770-96-2
H4AlNa
54
MFCD00192443
MNYCIYRTKITUKR-UHFFFAOYSA-N
sodium alumanuide,sodium aluminum hydride,aluminum sodium hydride,aluminumsodiumhydride,naalh4,sah22,sodiumtetrahydroaluminate,sodiumaluminumtetrahydride,na alh4,sodium tetrahydroaluminate
26266
CHEBI:30141
sodium;alumanuide
[Na+].[AlH4-]
13770-96-2
H4AlNa
54
MFCD00192443
MNYCIYRTKITUKR-UHFFFAOYSA-N
sodium alumanuide,sodium aluminum hydride,aluminum sodium hydride,aluminumsodiumhydride,naalh4,sah22,sodiumtetrahydroaluminate,sodiumaluminumtetrahydride,na alh4,sodium tetrahydroaluminate
26266
CHEBI:30141
sodium;alumanuide
[Na+].[AlH4-]
Assay Percent Range 93%
Solubility Information Solubility in water: reacts violently
Formula Weight 54
Percent Purity 93%
Packaging Glass bottle
Solubility Solubility in water: reacts violently.
Melting Point 178.0°C
Quantity 25g
Chemical Name or Material Sodium aluminium hydride