Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Aluminum Phosphate, Acidic, FCC, 95%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10305-76-7
10305-76-7

S1106, 10305-76-7, Na3Al3H14(PO4)8•4H2O

Spectrum™ Sodium Aluminum Phosphate, Acidic, FCC is used as an acid for baking powders for the chemical leavening of baked goods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 95%
Fluoride 2.5e-005
Lead (Pb) 2 mg/kg
Loss on Ignition 19.5-21%
Packaging Fiber Drum
Quantity 45 kg