Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium amide, 95%, pure
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 400°C |
| Melting Point | 210°C |
| Color | Gray-White to White |
| Physical Form | Powder, Irregular Flakes or Granules |
| Quantity | 2.5 kg |
| Assay Percent Range | 95% |
| Molecular Formula | H2NNa |
| Linear Formula | NaNH2 |
| Fieser | 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288 |
| Merck Index | 15,8714 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Very toxic to aquatic life. Reacts violently with water. May form explosive peroxides.< |
| Precautionary Statement | GHS P Statement Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present |
| EINECSNumber | 231-971- |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction