Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium amide, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
25g,25g
100g,100g
Packaging:
Glass bottle,Glass bottle
7782-92-5
H2NNa
39
MFCD00011117
ODZPKZBBUMBTMG-UHFFFAOYSA-N
sodium amide,sodamide,sodium amide na nh2,unii-5db3g6px9d,5db3g6px9d,sodium azanide,sodium amide, 50 wt. % suspension in toluene,sodiumamide,natriumamid,sodioamine
24533
sodium;azanide
[NH2-].[Na+]
7782-92-5
H2NNa
39
MFCD00011117
ODZPKZBBUMBTMG-UHFFFAOYSA-N
sodium amide,sodamide,sodium amide na nh2,unii-5db3g6px9d,5db3g6px9d,sodium azanide,sodium amide, 50 wt. % suspension in toluene,sodiumamide,natriumamid,sodioamine
24533
sodium;azanide
[NH2-].[Na+]
Quantity 25g,25g
Melting Point 210.0°C,210.0°C
Peroxides 0.1% max. (as NaNH2O2)
Packaging Glass bottle,Glass bottle
Boiling Point 400.0°C,400.0°C
Color Gray-White to White,Gray-White to White
Assay Percent Range 99%
Linear Formula NaNH2
Fieser 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
Merck Index 15, 8714
Solubility Information Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
Formula Weight 39
Physical Form Powder, Irregular Flakes or Granules
Percent Purity 99%
Grade Extra Pure
Free Alkali Passes Test (a small amount dropped into water, does not catch fire)
Solubility Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
Chemical Name or Material Sodium amide, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Very toxic to aquatic life.
Reacts violently with water.
May form explosive peroxides.<

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger

EINECSNumber : 231-971-0

TSCA : TSCA