Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ascorbate, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
134-03-2
C6H7NaO6
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
134-03-2
C6H7NaO6
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

SO108, 134-03-2, C6H7NaO6

Spectrum™ Sodium Ascorbate, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Specific Rotation +103° to +108°
Quantity 50 kg
Loss on Drying 0.0025
pH 7.0 to 8.0
CAS Max % 1
Percent Purity 99 to 101%
Grade USP