Learn More
BioVision IncSupplier Diversity Partner Sodium Assay Kit (Colorimetric): Sodium Assay Kit (Colorimetric): Convenient & Sensitive Colorimetric Assay for Quantitating sodium in a Variety of Biological Fluids such as Serum, urine, saliva etc. Detection Limit: as low as 25 µM. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K391100
Sodium Assay Kit (Colorimetric): Sodium (Na) is one of the most important electrolytes along with chloride, calcium and potassium. Healthy individuals can absorb sodium ingested in food, and kidneys maintain proper sodium balance by excreting its excess in urine. Hyponatremia (low sodium concentration in blood) can occur in patients with nephrotic syndrome, excessive vomiting and diarrhea, while Hypernatremia (high sodium concentration in blood) is developed in patients suffering from liver diseases, burns, and pregnancy. BioVision's Sodium Assay Kit offers a simple, two-step colorimetric assay that is based on the requirement of sodium ion as a cofactor for the enzymatic activity of ?-Galactosidase (?G). Endogenous mono-, di-, and trivalent ions, ascorbic acid, creatinine, glucose, urea, and bilirubin do not interfere with the assay. The kit can detect Sodium concentration as low as 25 ¿M in a variety of samples. BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9734