Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sodium Azide, ≥99%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Plastic bottle
26628-22-8
N3Na
65.011
MFCD00003536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
26628-22-8
N3Na
65.011
MFCD00003536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
UN Number UN1687
Formula Weight 65.01
Physical Form Crystalline Powder
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 1.846
Melting Point ~275°C (decomposition)
Quantity 500g
Chemical Name or Material Sodium Azide