Learn More
Sodium azide, 99%, Thermo Scientific Chemicals
Description
In airbag inflation, as a preservative in diagnostic medicinals.Sodium azide is used in organic synthesis for the introduction of an azide group to prepare a desired compound. It acts as a precursor to prepare inorganic azides like lead azide and silver azide. In biochemistry, it is useful as a probe reagent and a preservative. It also acts as a nitridizing agent and cytochrome oxidase inhibitor. Further, it is used as a propellant in airbags.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∽275° (decomposition) |
Physical Form | Powder or Granules |
Quantity | 10 g |
Solubility Information | Soluble in water and liquid ammonia. Slightly soluble in alcohol. Insoluble in ether. |
Formula Weight | 65.01 |
Sensitivity | Air Sensitive |
Density | 1.846 |
Chemical Name or Material | Sodium Azide |
Safety and Handling
Hazard Category | H300+H310+H330-H373 |
Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: II |
EINECSNumber | 247-852-1 |
RTECSNumber | VY8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.