Sodium Azide, 99%, Alfa Aesar

Manufacturer: Alfa Aesar 1431409

Quantity 10g

 View more versions of this product

Catalog No. AA1431409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26628-22-8
    Molecular Formula NaN3
    Formula Weight 65.01
    Chemical Name or Material Sodium Azide

    Specifications

    Formula Weight 65.01
    Quantity 10g
    Molecular Formula NaN3
    CAS 26628-22-8
    Chemical Name or Material Sodium Azide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit