Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium azide, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
26628-22-8
N3Na
65.01
MFCD00003536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
26628-22-8
N3Na
65.01
MFCD00003536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
C-X bond formation reagent, For Azidation/Diazo transfer
Additional Information sodium hydroxide: 0.1% max.
Assay Percent Range 98.5% min
Linear Formula NaN3
Fieser 01,1041; 02,376; 03,259; 04,440; 05,593; 15,289; 17,68
Merck Index 15, 8717
Solubility Information Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
Formula Weight 65.01
Physical Form Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Density 1.8500g/mL
Heavy Metals (as Pb) 20ppm max.
Insoluble in Water 0.05% max.
Insoluble Matter 0.05% max. (in water)
Loss on Drying 0.5% max.
Packaging Plastic bottle
Sodium Carbonate 0.15% max.
Solubility Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
Specific Gravity 1.85
Decomposition Information >275°C
Boiling Point 300.0°C
Color White
Melting Point 275.0°C
pH 9 (5% min. soln.)
Quantity 500g
Chemical Name or Material Sodium azide, 99%
Show More Show Less

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 247-852-1

RTECSNumber : VY8050000

TSCA : TSCA