Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Azide OR, Macron Fine Chemicals™

Click to view available options
Quantity:
500g
5kg
Packaging:
Poly
Poly Pail
26628-22-8
N3Na
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
26628-22-8
N3Na
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
Formula Weight 65.01
Packaging Poly Pail
Quantity 5kg
Chemical Name or Material Sodium Azide