Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Benzoate, N.F. - F.C.C., J.T.Baker™
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Formula Weight | 144.11 |
Grade | FCC/NF |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.44kg |
Quantity | 2.5kg |
Chemical Name or Material | Sodium Benzoate |