Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium bicarbonate, 1M buffer soln., pH 9.4

Click to view available options
Quantity:
250mL
500mL
144-55-8
CHNaO3
84.006
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]
144-55-8
CHNaO3
84.006
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]
Applications
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
Concentration 1M buffer soln.
Physical Form Liquid
pH 9.4
Quantity 500mL
Chemical Name or Material Sodium bicarbonate buffer soln.

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA