Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, 99+%, for HPLC, Acros Organics

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
144-55-8
CHNaO3
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]
144-55-8
CHNaO3
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]
For HPLC
Assay Percent Range 99+%
Linear Formula NaHCO3
Merck Index 15,8719
Solubility Information Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol
Formula Weight 84.01
Percent Purity 99+%
Grade HPLC
Packaging Glass bottle
Solubility Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
Melting Point 270.0°C
Quantity 1kg
Chemical Name or Material Sodium bicarbonate
Show More Show Less

EINECSNumber : 205-633-8