Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, 99+%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
516892
CHEBI:32139
sodium;hydrogen carbonate
C(=O)(O)[O-].[Na+]

Sodium bicarbonate, 99+%, CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 5kg, 20ppm max., 100ppm max., CHEBI:32139, White, 205-633-8

Sodium bicarbonate, 99+%, CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 5kg, 20ppm max., 100ppm max., CHEBI:32139, White, 205-633-8

Assay Percent Range 99+%
Linear Formula NaHCO3
Ammonium 20ppm max.
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15, 8719
Solubility Information Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol
Formula Weight 84.01
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Calcium (Ca) 100ppm max.
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Insoluble Matter 0.015% max
Iron (Fe) 10ppm max.
Packaging Plastic bottle
Potassium (K) 100ppm max.
Solubility Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
Sulfate 100ppm max.
Color White
Melting Point 270.0°C
Quantity 1kg
Chemical Name or Material Sodium bicarbonate
Show More Show Less

EINECSNumber : 205-633-8