Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Bicarbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Specifications
Formula Weight | 84.01 |
Physical Form | Powder |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.20kg |
Quantity | 2.5kg |
Chemical Name or Material | Sodium Bicarbonate |