Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

144-55-8
CHNaO3
84.006
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
CHEBI:32139
CAS 144-55-8
Molecular Formula CHNaO3
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES C(=O)(O)[O-].[Na+]
IUPAC Name sodium;hydrogen carbonate
Molecular Weight (g/mol) 84.006
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.20kg
Quantity 2.5kg
Chemical Name or Material Sodium Bicarbonate
Grade FCC/USP
Show More Show Less