Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, USP, EP, BP, JP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
144-55-8
CHNaO3
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
[Na+].OC([O-])=O
144-55-8
CHNaO3
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
[Na+].OC([O-])=O

SO125, 144-55-8, NaHCO3

Spectrum™ Sodium Bicarbonate, Powder, USP, EP, BP, JP is used as an antacid to treat heartburn, indigestion, and upset stomach. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Arsenic (As) 2 ppm
Percent Purity 99 to 100.5%
Calcium (Ca) 100 ppm
Iron (Fe) 20 ppm
Loss on Drying 0.0025
Normal Carbonate To pass test
Packaging Poly Bottle
Sulfur Compounds 0.00015
pH 7.9 to 8.4
Quantity 6 x 500 g
Show More Show Less