Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bis(2-methoxyethoxy)aluminumhydride, 70 wt. % Sol. in Toluene (ca. 3.5M), ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
22722-98-1,108-88-3
C6H16AlNaO4
22722-98-1,108-88-3
C6H16AlNaO4
Reduction reagent
Linear Formula [(CH3OCH2CH2O)2AlH2]Na
Fieser 02,382; 03,260; 04,441; 05,596; 06,528; 07,327; 08,448; 12,440; 15,290
Density 1.0360g/mL
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Formula Weight 202.16
Physical Form Viscous Liquid
Percent Purity 68.5 to 71.5%
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Specific Gravity 1.036
Viscosity 65 mPa.s (20°C)
Boiling Point 110.0°C
Color Amber to Colorless
Quantity 50g
Flash Point 4°C
Chemical Name or Material Sodium bis(2-methoxyethoxy)aluminium hydride
Show More Show Less