Alfa Aesar™ Sodium bis(trimethylsilyl)amide, 98%

Manufacturer: Alfa Aesar™ 3350518

Quantity 50g

 View more versions of this product

Catalog No. AA3350518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1070-89-9
    Linear Formula (CH3)3SiNNaSi(CH3)3
    Formula Weight 183.38
    MDL Number MFCD00009835
    UN Number UN3096
    Synonym Sodium hexamethyldisilazane; N-Sodiohexamethyldisilazane
    Chemical Name or Material Sodium bis(trimethylsilyl)amide

    Specifications

    Formula Weight 183.38
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310-P370+P378c-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    F exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use ... for extinction.
    Store in a dry place.
    Physical Form Crystalline
    MDL Number MFCD00009835
    CAS 1070-89-9
    UN Number UN3096
    Synonym Sodium hexamethyldisilazane; N-Sodiohexamethyldisilazane
    Color White
    Quantity 50g
    Odor Odorless
    TSCA TSCA
    Melting Point 171° to 175°C
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, WATER-REACTIVE, N.O.S.
    Linear Formula (CH3)3SiNNaSi(CH3)3
    Solubility Information Soluble in hexane, toluene, ether
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 1 Danger
    Chemical Name or Material Sodium bis(trimethylsilyl)amide
    Assay 98%

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310-P370+P378c-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    F exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use ... for extinction.
    Store in a dry place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit