Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bismuthate, ACS reagent, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
12232-99-4
BiNaO3
279.96
PNYYBUOBTVHFDN-UHFFFAOYSA-N
sodium bismuthate,bismuth sodium oxide,sodium bismuthate v,bismuth sodium trioxide,bismuthate bio31-, sodium,sodium oxido dioxo bismuth,sodium bismuth oxide,bio3.na,bismuthate bio31#-, sodium
4063671
sodium;oxido(dioxo)bismuth
[O-][Bi](=O)=O.[Na+]
12232-99-4
BiNaO3
279.96
PNYYBUOBTVHFDN-UHFFFAOYSA-N
sodium bismuthate,bismuth sodium oxide,sodium bismuthate v,bismuth sodium trioxide,bismuthate bio31-, sodium,sodium oxido dioxo bismuth,sodium bismuth oxide,bio3.na,bismuthate bio31#-, sodium
4063671
sodium;oxido(dioxo)bismuth
[O-][Bi](=O)=O.[Na+]
Linear Formula NaBiO3
Merck Index 15,8721
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids
Formula Weight 279.96
Grade ACS Reagent
Chloride 0.002% max
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids
Quantity 100g
Chemical Name or Material Sodium bismuthate

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/a

GHS Signal Word: Warning

EINECSNumber : 235-455-6