Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium bismuthate(V), 85%, ACROS Organics™
Click to view available options
Quantity:
25g
250g
1kg
Packaging:
Glass bottle
Specifications
Assay Percent Range | 80% min. (Iodometry) |
Linear Formula | NaBiO3 |
Merck Index | 15,8721 |
Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids |
Formula Weight | 279.96 |
Physical Form | Powder |
Percent Purity | 85% |
Chloride | 20ppm max |
Chlorine (Cl) | 20ppm max. |
Manganese (Mn) | 5ppm max. |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/a
GHS Signal Word: Warning
EINECSNumber : 235-455-6