Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bisulfate monohydrate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
10034-88-5
HNaO4S·H2O
138.08
JXHZRQHZVYDRGX-UHFFFAOYSA-M
sodium bisulfate monohydrate,sodium hydrogen sulfate monohydrate,unii-3kls9y77yj,sulfuric acid, monosodium salt, hydrate,3kls9y77yj,sodium hydrogensulfate hydrate,sulfuric acid, monosodium salt, monohydrate,sodium hydrate hydrogen sulfate,sodium hydrogen sulfate-1-hydrate,acmc-20ajox
23673662
sodium;hydrogen sulfate;hydrate
O.OS(=O)(=O)[O-].[Na+]
10034-88-5
HNaO4S·H2O
138.08
JXHZRQHZVYDRGX-UHFFFAOYSA-M
sodium bisulfate monohydrate,sodium hydrogen sulfate monohydrate,unii-3kls9y77yj,sulfuric acid, monosodium salt, hydrate,3kls9y77yj,sodium hydrogensulfate hydrate,sulfuric acid, monosodium salt, monohydrate,sodium hydrate hydrogen sulfate,sodium hydrogen sulfate-1-hydrate,acmc-20ajox
23673662
sodium;hydrogen sulfate;hydrate
O.OS(=O)(=O)[O-].[Na+]
Assay Percent Range 99%
Linear Formula NaHSO4·H2O
Merck Index 15,8722
Solubility Information Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and,h2so4
Formula Weight 138.08
Percent Purity 99%
Grade Analysis
Packaging Plastic Bottle
Solubility Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4
Melting Point 58.0°C
Quantity 2.5kg
Chemical Name or Material Sodium bisulfate monohydrate
Show More Show Less