Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bitartrate, Monohydrate, Powder, Reagent, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
6131-98-2
C4H7NaO7
190.08
LLVQEXSQFBTIRD-UHFFFAOYNA-M
sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
O.[Na+].OC(C(O)C([O-])=O)C(O)=O
6131-98-2
C4H7NaO7
190.08
LLVQEXSQFBTIRD-UHFFFAOYNA-M
sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
O.[Na+].OC(C(O)C([O-])=O)C(O)=O

S1175, 6131-98-2, NaHC4H4O6•H2O

Spectrum™ Sodium Bitartrate, Monohydrate, Powder, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Grade Reagent
CAS Max % 1
Percent Purity 99%
Heavy Metals (as Pb) 2e-005
Iron (Fe) 3e-005
Packaging Poly Bottle
Quantity 500 g