Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate Decahydrate, Technical, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1303-96-4
B4H20Na2O17
381.36
OJPFLODMGUZMKD-UHFFFAOYSA-N
disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
1303-96-4
B4H20Na2O17
381.36
OJPFLODMGUZMKD-UHFFFAOYSA-N
disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

S1186, 1303-96-4, Na2B4O7•10H2O

Spectrum™ Sodium Borate Decahydrate, Technical is also known simply as Borax and is a mineral that occurs naturally in deposits of the mineral Evaporite. Its uses vary from household products, such as hand soap and water softening agents, to use as a flux in welding and can also be found in small scale gold mining operations as a substitute to mercury.

Grade Technical
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg