Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borodeuteride, 98 atom% D, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
15681-89-7
BH4Na
41.856
MFCD00003519
YOQDYZUWIQVZSF-XWFVQAFUSA-N
sodium borodeuteride,sodium tetradeuterioboron,sodium borohydride-d4,sodium 2h4 tetrahydroborate 1-,sulfatase,borate 1-, tetrahydro-d4-, sodium,nabd4,sodium 2h4 boranuide,deuterated sodium borohydride
23673181
sodium;tetradeuterioboranuide
[BH4-].[Na+]
15681-89-7
BH4Na
41.856
MFCD00003519
YOQDYZUWIQVZSF-XWFVQAFUSA-N
sodium borodeuteride,sodium tetradeuterioboron,sodium borohydride-d4,sodium 2h4 tetrahydroborate 1-,sulfatase,borate 1-, tetrahydro-d4-, sodium,nabd4,sodium 2h4 boranuide,deuterated sodium borohydride
23673181
sodium;tetradeuterioboranuide
[BH4-].[Na+]
Color Gray to White
Quantity 1g
Packaging Glass bottle
Deuterium (D) 97.5% min.
Assay Percent Range 98 atom% D
Melting Point >300.0°C
Linear Formula NaBD4
Fieser 03,281
Formula Weight 41.86
Physical Form Powder and/or Chunks
Percent Purity 98 atom% D
Grade NMR
Chemical Name or Material Sodium borodeuteride, 98 atom % D
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 239-764-7