Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borohydride, 98+%, powder, ACROS Organics™

Click to view available options
Quantity:
5g
10g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
16940-66-2
H4BNa
MFCD00003518
Sodium tetrahydroborate,SBH
16940-66-2
H4BNa
MFCD00003518
Sodium tetrahydroborate,SBH
Reduction reagent
Additional Information Vapor Pressure: Negligible
Assay Percent Range 98+%
Linear Formula NaBH4
Fieser 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Merck Index 15, 8727
Formula Weight 37.83
Physical Form Powder
Percent Purity ≥98%
Density 1.3
Packaging Plastic drum
Solubility Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
Decomposition Information >220°C
Color White
Melting Point 360.0°C
Quantity 2.5kg
Chemical Name or Material Sodium borohydride
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger

EINECSNumber : 241-004-4

RTECSNumber : ED3325000

TSCA : TSCA