Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, 98+% (Sodium tetrahydridoborate), Reagents

Catalog No. p-200021151
Click to view available options
Quantity:
10 g
25 g
100 g

Sodium Borohydride, 98+% (Sodium tetrahydridoborate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium Tetrahydroborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point 400°C
Melting Point 400°C
Color White to Gray
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.074
Chemical Name or Material Sodium Borohydride, 98+% (Sodium tetrahydridoborate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.