Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borohydride, 99%, (VenPure(TM) SF granules), granules, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
16940-66-2
H4BNa
37.83
Sodium tetrahydroborate,SBH
16940-66-2
H4BNa
37.83
Sodium tetrahydroborate,SBH
Assay Percent Range 98.5% min. (Hydrogen evolution)
Linear Formula NaBH4
Merck Index 15,8727
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ammonia- reacts with acids,soluble in amines,pyridine and dimethylether of,diethylene glycol,insol.in other ethers,hydrocarb.,alkyl chlorides,reacts with oxidizing agents
Formula Weight 37.83
Percent Purity 99%
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
Melting Point 360.0°C
Quantity 500g
Chemical Name or Material Sodium borohydride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf

GHS Signal Word: Danger

EINECSNumber : 241-004-4