Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Acros Organics

Click to view available options
Quantity:
100mL
250mL
1L
2.5L
Packaging:
Plastic bottle
16940-66-2,1310-73-2,67-56-1,7732-18-5
BH4Na
37.82
MFCD00003518
Sodium tetrahydroborate,SBH
16940-66-2,1310-73-2,67-56-1,7732-18-5
BH4Na
37.82
MFCD00003518
Sodium tetrahydroborate,SBH
Reduction reagent
Linear Formula NaBH4
Merck Index 15, 8727
Solubility Information Solubility in water: completely soluble
Formula Weight 37.82
Physical Form Liquid
Percent Purity 11 to 13%
Density 1.4000g/mL
Packaging Plastic bottle
Solubility Solubility in water: completely soluble.
Specific Gravity 1.4
Viscosity 60.43 mPa.s (20°C)
Boiling Point 130.0°C to 135.0°C
Color Colorless
Quantity 250mL
Chemical Name or Material Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce

GHS Signal Word: Danger

EINECSNumber : 241-004-4