Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7789-38-0
BrNaO3
150.9
MFCD00003476
XUXNAKZDHHEHPC-UHFFFAOYSA-M
sodium bromate,bromic acid, sodium salt,dyetone,bromate de sodium,sodium bromate dot,neutralizer k-126,neutralizer k-140,neutralizer k-938,sodium bromate nabro3,bromate de sodium french
23668195
CHEBI:75229
sodium;bromate
[O-]Br(=O)=O.[Na+]
7789-38-0
BrNaO3
150.9
MFCD00003476
XUXNAKZDHHEHPC-UHFFFAOYSA-M
sodium bromate,bromic acid, sodium salt,dyetone,bromate de sodium,sodium bromate dot,neutralizer k-126,neutralizer k-140,neutralizer k-938,sodium bromate nabro3,bromate de sodium french
23668195
CHEBI:75229
sodium;bromate
[O-]Br(=O)=O.[Na+]
Oxidation reagent
Assay Percent Range 99+%
Linear Formula NaBrO3
Fieser 01,1055
Merck Index 15,8728
Solubility Information Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol ,ether and aceton
Formula Weight 150.9
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Bromine (Br) 500ppm max.
Density 3.3300g/mL
Heavy Metals (as Pb) 50ppm max.
Iron (Fe) 50ppm max.
Packaging Glass bottle
Solubility Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol , ether and aceton
Specific Gravity 3.33
Sulfate 500ppm max.
Color White
Melting Point 381.0°C
Quantity 2.5kg
Chemical Name or Material Sodium bromate, 99+%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut

GHS Signal Word: Danger

EINECSNumber : 232-160-4

RTECSNumber : EF8750000

TSCA : TSCA