Sodium bromide, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 1403736

Quantity 500g

 View more versions of this product

Catalog No. AA1403736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-15-6
    Physical Form Granular
    Assay Percent Range 97%
    Molecular Formula NaBr
    Formula Weight 102.89
    MDL Number MFCD00003475
    Chemical Name or Material Sodium bromide

    Specifications

    Formula Weight 102.89
    Quantity 500g
    EINECS Number 231-599-9
    Physical Form Granular
    Density 3.203g/mL
    Assay Percent Range 97%
    Molecular Formula NaBr
    MDL Number MFCD00003475
    CAS 7647-15-6
    Chemical Name or Material Sodium bromide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit