Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromide, 99+%, extra pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
5kg
Packaging:
Plastic bottle
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]
C-X bond formation reagent
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15, 8729
Arsenic (As) 2ppm max.
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C),1210g/L water (100°C),1g/6 mL methanol,7g/100 mL 95% ethanol
Barium (Ba) 20ppm max.
Formula Weight 102.89
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Bromate 10ppm max.
Calcium (Ca) 50ppm max.
Chlorine (Cl) 2000ppm max.
Heavy Metals (as Pb) 10ppm max.
Iodine (I) 200ppm max.
Iron (Fe) 10ppm max.
Loss on Drying 1% max. (120°C)
Magnesium (Mg) 10ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
Sulfate 50ppm max.
Boiling Point 1390.0°C
Color White
Melting Point 755.0°C
pH 5 to 7 (5% soln. at 20°C)
Quantity 5kg
Chemical Name or Material Sodium bromide, Anhydrous
Show More Show Less

EINECSNumber : 231-599-9