Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromide, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15,8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C),1210g/L water (100°C),1g/6 mL methanol,7g/100 mL 95% ethanol
Formula Weight 102.89
Grade ACS Reagent
Chloride 0.2% max
Insoluble Matter 0.005% max
Packaging Glass bottle
Solubility Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
Sulfate 0.002% max
Boiling Point 1390.0°C
Melting Point 755.0°C
Quantity 100g
Chemical Name or Material Sodium bromide
Show More Show Less

EINECSNumber : 231-599-9