Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bromide, ACS Reagent, Reagents

Catalog No. 01337750
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Catalog No. Quantity
01-337-750 12 kg
01-337-749 2.5 kg
01-337-748 500 g
3 options

Catalog No. 01-337-750

Supplier: Reagents Holdings Llc C131100012KG

Only null left
Add to Cart
Add to Cart

Sodium Bromide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 3.21
Chemical Name or Material Sodium Bromide, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.